Assay Details
| Gene Symbol: Usp14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273392.1 | NP_001260321.1 | 6 - 7 | 984 | 64 | |
| NM_135533.2 | NP_609377.1 | 6 - 7 | 958 | 64 | ||
| GenBank mRNA | BT031279.1 | - | 6 - 7 | 919 | 64 | |
More Information
Set Membership: |
|

Usp14
Genomic Map