Assay Details
| Gene Symbol: Klp31E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169464.1 | NP_001162935.1 | 5 - 6 | 1541 | 99 | |
| NM_001169465.2 | NP_001162936.1 | 6 - 7 | 1642 | 99 | ||
| NM_001169466.2 | NP_001162937.2 | 6 - 7 | 1662 | 99 | ||
| NM_135554.3 | NP_609398.2 | 6 - 7 | 1662 | 99 | ||
| GenBank mRNA | BT001471.1 | - | 6 - 7 | 1645 | 99 | |
More Information
Set Membership: |
|

Klp31E
Genomic Map