Assay Details
| Gene Symbol: Nse4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169467.1 | NP_001162938.1 | 1 - 2 | 177 | 72 | |
| NM_135561.3 | NP_609405.2 | 1 - 2 | 177 | 72 | ||
| GenBank mRNA | BT023592.1 | - | 1 - 2 | 177 | 72 | |
More Information
Set Membership: |
|

Nse4
Genomic Map