Assay Details
| Gene Symbol: Porin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273435.1 | NP_001260364.1 | 2 - 3 | 411 | 64 | |
| NM_135618.2 | NP_609462.1 | 1 - 2 | 470 | 64 | ||
| GenBank mRNA | AJ580916.1 | - | 1 - 2 | 324 | 64 | |
| AY118823.1 | - | 1 - 2 | 448 | 64 | ||
More Information
Set Membership: |
|

Porin2
Genomic Map