Assay Details
| Gene Symbol: YL-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298894.1 | NP_001285823.1 | 1 - 2 | 246 | 70 | |
| NM_135631.4 | NP_609475.1 | 1 - 2 | 246 | 70 | ||
More Information
Set Membership: |
|

YL-1
Genomic Map