Assay Details
| Gene Symbol: YL-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298894.1 | NP_001285823.1 | 3 - 4 | 682 | 76 | |
| NM_135631.4 | NP_609475.1 | 3 - 4 | 682 | 76 | ||
| GenBank mRNA | AY122237.1 | - | 3 - 4 | 670 | 76 | |
More Information
Set Membership: |
|

YL-1
Genomic Map