Assay Details
| Gene Symbol: Cog8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135659.3 | NP_609503.2 | 2 - 3 | 613 | 64 | |
| GenBank mRNA | BT031276.1 | - | 2 - 3 | 579 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cog8| Gene Symbol: Cog8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135659.3 | NP_609503.2 | 2 - 3 | 613 | 64 | |
| GenBank mRNA | BT031276.1 | - | 2 - 3 | 579 | 64 | |
Set Membership: |
|