Assay Details
Gene Symbol: dl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169529.1 | NP_001163000.1 | 2 - 3 | 675 | 67 | |
NM_001169530.1 | NP_001163001.1 | 2 - 3 | 589 | 67 | ||
NM_001299085.1 | NP_001286014.1 | 2 - 3 | 675 | 67 | ||
NM_165217.3 | NP_724052.1 | 3 - 4 | 574 | 67 | ||
NM_165218.3 | NP_724053.1 | 3 - 4 | 918 | 67 | ||
NM_165219.2 | NP_724054.1 | 3 - 4 | 918 | 67 | ||
GenBank mRNA | BT099643.1 | - | 2 - 3 | 662 | 67 | |
BT133461.1 | - | 3 - 4 | 570 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
