Assay Details
Gene Symbol: dl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169529.1 | NP_001163000.1 | 3 - 4 | 858 | 68 | |
NM_001169530.1 | NP_001163001.1 | 3 - 4 | 772 | 68 | ||
NM_001299085.1 | NP_001286014.1 | 3 - 4 | 858 | 68 | ||
NM_165217.3 | NP_724052.1 | 4 - 5 | 757 | 68 | ||
NM_165218.3 | NP_724053.1 | 4 - 5 | 1101 | 68 | ||
NM_165219.2 | NP_724054.1 | 4 - 5 | 1101 | 68 | ||
GenBank mRNA | BT099643.1 | - | 3 - 4 | 845 | 68 | |
BT133461.1 | - | 4 - 5 | 753 | 68 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
