Assay Details
Gene Symbol: dl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169529.1 | NP_001163000.1 | 5 - 6 | 1330 | 65 | |
NM_001169530.1 | NP_001163001.1 | 5 - 6 | 1244 | 65 | ||
NM_165217.3 | NP_724052.1 | 6 - 7 | 1229 | 65 | ||
NM_165218.3 | NP_724053.1 | 6 - 7 | 1573 | 65 | ||
GenBank mRNA | BT133461.1 | - | 6 - 7 | 1225 | 65 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
