Assay Details
Gene Symbol: dl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169529.1 | NP_001163000.1 | 6 - 7 | 1438 | 79 | |
NM_001169530.1 | NP_001163001.1 | 6 - 7 | 1352 | 79 | ||
NM_165217.3 | NP_724052.1 | 7 - 8 | 1337 | 79 | ||
NM_165218.3 | NP_724053.1 | 7 - 8 | 1681 | 79 | ||
GenBank mRNA | BT133461.1 | - | 7 - 8 | 1333 | 79 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
