Assay Details
Gene Symbol: BicD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259128.2 | NP_001246057.1 | 1 - 2 | 1122 | 72 | |
NM_001273601.1 | NP_001260530.1 | 1 - 2 | 1122 | 72 | ||
NM_001273602.1 | NP_001260531.1 | 1 - 2 | 1122 | 72 | ||
NM_165220.3 | NP_724056.1 | 1 - 2 | 1122 | 72 | ||
GenBank mRNA | AY069452.1 | - | 1 - 2 | 625 | 72 | |
X51652.1 | - | 1165 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
