Assay Details
Gene Symbol: BicD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273601.1 | NP_001260530.1 | 7 - 8 | 3120 | 102 | |
NM_001273602.1 | NP_001260531.1 | 7 - 8 | 3120 | 102 | ||
NM_165220.3 | NP_724056.1 | 7 - 8 | 3120 | 102 | ||
GenBank mRNA | AY069452.1 | - | 7 - 8 | 2623 | 102 | |
X51652.1 | - | 3163 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
