Assay Details
Gene Symbol: fws | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259130.2 | NP_001246059.1 | 2 - 3 | 676 | 88 | |
NM_001299086.1 | NP_001286015.1 | 2 - 3 | 650 | 88 | ||
NM_001299087.1 | NP_001286016.1 | 1 - 2 | 699 | 88 | ||
NM_078866.4 | NP_523590.1 | 1 - 2 | 729 | 88 | ||
NM_165221.3 | NP_724058.1 | 2 - 3 | 680 | 88 | ||
GenBank mRNA | BT003257.1 | - | 1 - 2 | 682 | 88 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
