Assay Details
Gene Symbol: CadN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032106.2 | NP_001027277.1 | 13 - 14 | 4951 | 65 | |
NM_001032107.3 | NP_001027278.1 | 13 - 14 | 4951 | 65 | ||
NM_001032108.2 | NP_001027279.1 | 13 - 14 | 4951 | 65 | ||
NM_001032109.3 | NP_001027280.1 | 13 - 14 | 4951 | 65 | ||
NM_165224.2 | NP_724068.1 | 12 - 13 | 4939 | 65 | ||
NM_165225.2 | NP_724069.1 | 12 - 13 | 4939 | 65 | ||
NM_165226.2 | NP_724070.1 | 12 - 13 | 4939 | 65 | ||
NM_165227.3 | NP_724071.1 | 14 - 15 | 5095 | 65 | ||
NM_165228.2 | NP_724072.1 | 12 - 13 | 4939 | 65 | ||
NM_165229.2 | NP_724073.1 | 12 - 13 | 4939 | 65 | ||
NM_165230.2 | NP_724074.1 | 12 - 13 | 4939 | 65 | ||
NM_165231.3 | NP_724075.1 | 12 - 13 | 4939 | 65 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
