Assay Details
| Gene Symbol: CadN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032108.2 | NP_001027279.1 | 14 - 15 | 5065 | 62 | |
| NM_001032109.3 | NP_001027280.1 | 14 - 15 | 5065 | 62 | ||
| NM_165224.2 | NP_724068.1 | 13 - 14 | 5053 | 62 | ||
| NM_165225.2 | NP_724069.1 | 13 - 14 | 5053 | 62 | ||
| NM_165228.2 | NP_724072.1 | 13 - 14 | 5053 | 62 | ||
| NM_165229.2 | NP_724073.1 | 13 - 14 | 5053 | 62 | ||
More Information
Set Membership: |
|

CadN
Genomic Map