Assay Details
| Gene Symbol: CadN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032106.2 | NP_001027277.1 | 16 - 17 | 5369 | 60 | |
| NM_001032107.3 | NP_001027278.1 | 16 - 17 | 5369 | 60 | ||
| NM_001032108.2 | NP_001027279.1 | 16 - 17 | 5366 | 60 | ||
| NM_001032109.3 | NP_001027280.1 | 16 - 17 | 5366 | 60 | ||
| NM_165224.2 | NP_724068.1 | 15 - 16 | 5354 | 60 | ||
| NM_165225.2 | NP_724069.1 | 15 - 16 | 5354 | 60 | ||
| NM_165226.2 | NP_724070.1 | 15 - 16 | 5357 | 60 | ||
| NM_165227.3 | NP_724071.1 | 17 - 18 | 5513 | 60 | ||
| NM_165228.2 | NP_724072.1 | 15 - 16 | 5354 | 60 | ||
| NM_165229.2 | NP_724073.1 | 15 - 16 | 5354 | 60 | ||
| NM_165230.2 | NP_724074.1 | 15 - 16 | 5357 | 60 | ||
| NM_165231.3 | NP_724075.1 | 15 - 16 | 5357 | 60 | ||
More Information
Set Membership: |
|

CadN
Genomic Map