Assay Details
Gene Symbol: CadN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032106.2 | NP_001027277.1 | 22 - 23 | 9782 | 92 | |
NM_001032107.3 | NP_001027278.1 | 22 - 23 | 9779 | 92 | ||
NM_001032108.2 | NP_001027279.1 | 22 - 23 | 9776 | 92 | ||
NM_001032109.3 | NP_001027280.1 | 22 - 23 | 9779 | 92 | ||
NM_165224.2 | NP_724068.1 | 21 - 22 | 9764 | 92 | ||
NM_165225.2 | NP_724069.1 | 21 - 22 | 9767 | 92 | ||
NM_165226.2 | NP_724070.1 | 21 - 22 | 9767 | 92 | ||
NM_165227.3 | NP_724071.1 | 23 - 24 | 9926 | 92 | ||
NM_165228.2 | NP_724072.1 | 21 - 22 | 9764 | 92 | ||
NM_165229.2 | NP_724073.1 | 21 - 22 | 9767 | 92 | ||
NM_165230.2 | NP_724074.1 | 21 - 22 | 9767 | 92 | ||
NM_165231.3 | NP_724075.1 | 21 - 22 | 9770 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
