Assay Details
Gene Symbol: CadN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032106.2 | NP_001027277.1 | 19 - 20 | 9166 | 69 | |
NM_001032109.3 | NP_001027280.1 | 19 - 20 | 9163 | 69 | ||
NM_165225.2 | NP_724069.1 | 18 - 19 | 9151 | 69 | ||
NM_165227.3 | NP_724071.1 | 20 - 21 | 9310 | 69 | ||
NM_165229.2 | NP_724073.1 | 18 - 19 | 9151 | 69 | ||
NM_165231.3 | NP_724075.1 | 18 - 19 | 9154 | 69 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
