Assay Details
| Gene Symbol: CadN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032106.2 | NP_001027277.1 | 14 - 15 | 5066 | 56 | |
| NM_001032107.3 | NP_001027278.1 | 14 - 15 | 5066 | 56 | ||
| NM_165226.2 | NP_724070.1 | 13 - 14 | 5054 | 56 | ||
| NM_165227.3 | NP_724071.1 | 15 - 16 | 5210 | 56 | ||
| NM_165230.2 | NP_724074.1 | 13 - 14 | 5054 | 56 | ||
| NM_165231.3 | NP_724075.1 | 13 - 14 | 5054 | 56 | ||
More Information
Set Membership: |
|

CadN
Genomic Map