Assay Details
| Gene Symbol: CadN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_165228.2 | NP_724072.1 | 7 - 8 | 4039 | 74 | |
| NM_165229.2 | NP_724073.1 | 7 - 8 | 4039 | 74 | ||
| NM_165230.2 | NP_724074.1 | 7 - 8 | 4039 | 74 | ||
| NM_165231.3 | NP_724075.1 | 7 - 8 | 4039 | 74 | ||
More Information
Set Membership: |
|

CadN
Genomic Map