Assay Details
| Gene Symbol: Socs36E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273605.2 | NP_001260534.1 | 3 - 4 | 1855 | 72 | |
| NM_078869.6 | NP_523593.5 | 3 - 4 | 2024 | 72 | ||
| NM_165243.3 | NP_724096.1 | 3 - 4 | 2024 | 72 | ||
| GenBank mRNA | AF435838.1 | - | 3 - 4 | 2018 | 72 | |
More Information
Set Membership: |
|

Socs36E
Genomic Map