Assay Details
Gene Symbol: RtGEF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103714.3 | NP_001097184.2 | 3 - 4 | 687 | 67 | |
NM_001103715.4 | NP_001097185.2 | 3 - 4 | 1047 | 67 | ||
NM_001169550.2 | NP_001163021.1 | 3 - 4 | 1047 | 67 | ||
NM_001273693.1 | NP_001260622.1 | 3 - 4 | 687 | 67 | ||
NM_001273694.1 | NP_001260623.1 | 3 - 4 | 1047 | 67 | ||
NM_001382006.1 | NP_001369133.1 | 3 - 4 | 687 | 67 | ||
NM_057826.5 | NP_477174.1 | 3 - 4 | 1047 | 67 | ||
NM_165327.3 | NP_724258.1 | 3 - 4 | 687 | 67 | ||
GenBank mRNA | BT082080.1 | - | 3 - 4 | 1025 | 67 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Molecular Function -



Gene Ontology Categories:
