Assay Details
| Gene Symbol: Nhe2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144375.3 | NP_001137847.1 | 11 - 12 | 2687 | 85 | |
| NM_001144379.3 | NP_001137851.1 | 11 - 12 | 2687 | 85 | ||
| NM_001259187.2 | NP_001246116.1 | 11 - 12 | 2687 | 85 | ||
| NM_001259188.2 | NP_001246117.1 | 11 - 12 | 2687 | 85 | ||
| NM_136244.5 | NP_610088.2 | 11 - 12 | 2687 | 85 | ||
| NM_165355.4 | NP_724311.1 | 11 - 12 | 2681 | 85 | ||
More Information
Set Membership: |
|

Nhe2
Genomic Map