Assay Details
Gene Symbol: Nhe2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144375.3 | NP_001137847.1 | 12 - 13 | 2831 | 62 | |
NM_001144377.3 | NP_001137849.1 | 12 - 13 | 2855 | 62 | ||
NM_001144379.3 | NP_001137851.1 | 12 - 13 | 2831 | 62 | ||
NM_001259187.2 | NP_001246116.1 | 12 - 13 | 2831 | 62 | ||
NM_001259188.2 | NP_001246117.1 | 12 - 13 | 2831 | 62 | ||
NM_001273732.1 | NP_001260661.1 | 12 - 13 | 2855 | 62 | ||
NM_001273733.1 | NP_001260662.1 | 12 - 13 | 2855 | 62 | ||
NM_136244.5 | NP_610088.2 | 12 - 13 | 2831 | 62 | ||
NM_165355.4 | NP_724311.1 | 12 - 13 | 2825 | 62 |
Gene Symbol: CG46307 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001347785.1 | NP_001334745.1 | 12 - 13 | 2855 | 62 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
