Assay Details
| Gene Symbol: Mondo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169561.2 | NP_001163032.1 | 4 - 5 | 2058 | 69 | |
| NM_001169562.2 | NP_001163033.1 | 5 - 6 | 1149 | 69 | ||
| NM_001169563.2 | NP_001163034.1 | 4 - 5 | 2058 | 69 | ||
| NM_001169564.2 | NP_001163035.1 | 5 - 6 | 1570 | 69 | ||
| NM_001169565.2 | NP_001163036.1 | 4 - 5 | 1497 | 69 | ||
| NM_001169566.2 | NP_001163037.1 | 5 - 6 | 1051 | 69 | ||
| NM_001169567.2 | NP_001163038.1 | 5 - 6 | 1053 | 69 | ||
| NM_001169568.2 | NP_001163039.1 | 5 - 6 | 1289 | 69 | ||
| NM_136261.3 | NP_610105.2 | 5 - 6 | 1497 | 69 | ||
| NM_165367.2 | NP_724326.1 | 5 - 6 | 1058 | 69 | ||
| NM_165368.1 | NP_724327.1 | 3 - 4 | 521 | 69 | ||
| GenBank mRNA | AF264754.1 | - | 4 - 5 | 993 | 69 | |
| AY051894.2 | - | 4 - 5 | 1976 | 69 | ||
| BT133442.1 | - | 3 - 4 | 521 | 69 | ||
More Information
Set Membership: |
|

Mondo
Genomic Map