Assay Details
| Gene Symbol: Ac3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136272.3 | NP_610116.2 | 16 - 17 | 3140 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ac3| Gene Symbol: Ac3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136272.3 | NP_610116.2 | 16 - 17 | 3140 | 71 | |
Set Membership: |
|