Assay Details
Gene Symbol: Ac3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014496.3 | NP_001014496.1 | 3 - 4 | 697 | 79 | |
NM_136272.3 | NP_610116.2 | 3 - 4 | 697 | 79 | ||
GenBank mRNA | AY061347.1 | - | 1 - 2 | 407 | 79 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
