Assay Details
| Gene Symbol: Ac3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014496.3 | NP_001014496.1 | 9 - 10 | 1745 | 98 | |
| NM_136272.3 | NP_610116.2 | 10 - 11 | 1857 | 98 | ||
| GenBank mRNA | AY061347.1 | - | 7 - 8 | 1455 | 98 | |
More Information
Set Membership: |
|

Ac3
Genomic Map