Assay Details
| Gene Symbol: eEF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080366.3 | NP_525105.2 | 2 - 3 | 300 | 66 | |
| NM_165394.3 | NP_724357.2 | 2 - 3 | 300 | 66 | ||
| NM_165395.2 | NP_724358.2 | 2 - 3 | 348 | 66 | ||
| GenBank mRNA | AY075481.1 | - | 2 - 3 | 336 | 66 | |
More Information
Set Membership: |
|

eEF2
Genomic Map