Assay Details
Gene Symbol: sxc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078896.3 | NP_523620.1 | 6 - 7 | 905 | 149 | |
NM_165426.2 | NP_724406.1 | 6 - 7 | 964 | 149 | ||
NM_165427.2 | NP_724407.1 | 5 - 6 | 1025 | 149 | ||
GenBank mRNA | AF217788.1 | - | 6 - 7 | 950 | 149 | |
BT032865.1 | - | 5 - 6 | 1006 | 149 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
