Assay Details
Gene Symbol: sxc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078896.3 | NP_523620.1 | 9 - 10 | 3012 | 82 | |
NM_165426.2 | NP_724406.1 | 9 - 10 | 3071 | 82 | ||
NM_165427.2 | NP_724407.1 | 8 - 9 | 3132 | 82 | ||
GenBank mRNA | AF217788.1 | - | 9 - 10 | 3057 | 82 | |
BT032865.1 | - | 8 - 9 | 3113 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
