Assay Details
Gene Symbol: Src42A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057501.4 | NP_476849.1 | 1 - 2 | 860 | 76 | |
GenBank mRNA | AY058652.1 | - | 1 - 2 | 824 | 76 | |
BT044565.1 | - | 2 - 3 | 728 | 76 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
