Assay Details
Gene Symbol: Src42A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057501.4 | NP_476849.1 | 6 - 7 | 1419 | 69 | |
NM_136347.2 | NP_610191.2 | 6 - 7 | 4505 | 69 | ||
GenBank mRNA | AY058652.1 | - | 6 - 7 | 1383 | 69 | |
BT044565.1 | - | 7 - 8 | 1287 | 69 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
