Assay Details
Gene Symbol: Src42A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057501.4 | NP_476849.1 | 7 - 8 | 1573 | 128 | |
NM_136347.2 | NP_610191.2 | 7 - 8 | 4659 | 128 | ||
GenBank mRNA | AY058652.1 | - | 7 - 8 | 1537 | 128 | |
BT044565.1 | - | 8 - 9 | 1441 | 128 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
