Assay Details
Gene Symbol: Pld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144138.2 | NP_001137610.1 | 7 - 8 | 3650 | 72 | |
NM_078903.4 | NP_523627.3 | 6 - 7 | 3329 | 72 | ||
NM_165469.4 | NP_724469.2 | 7 - 8 | 3491 | 72 | ||
NM_165470.3 | NP_724470.2 | 6 - 7 | 3979 | 72 | ||
NM_165471.3 | NP_724471.2 | 6 - 7 | 3703 | 72 | ||
NM_165472.3 | NP_724472.3 | 6 - 7 | 3275 | 72 | ||
GenBank mRNA | AF145640.2 | - | 6 - 7 | 3941 | 72 | |
BT046138.1 | - | 6 - 7 | 3357 | 72 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
