Assay Details
| Gene Symbol: Tdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_165483.3 | NP_724489.1 | 2 - 3 | 297 | 85 | |
| GenBank mRNA | BT083414.1 | - | 2 - 3 | 281 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tdc2| Gene Symbol: Tdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_165483.3 | NP_724489.1 | 2 - 3 | 297 | 85 | |
| GenBank mRNA | BT083414.1 | - | 2 - 3 | 281 | 85 | |
Set Membership: |
|