Assay Details
| Gene Symbol: Tdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_165483.3 | NP_724489.1 | 6 - 7 | 1376 | 60 | |
| GenBank mRNA | BT083414.1 | - | 6 - 7 | 1360 | 60 | |
More Information
Set Membership: |
|

Tdc2
Genomic Map