Assay Details
Gene Symbol: pk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_165508.2 | NP_724534.1 | 4 - 5 | 1663 | 66 | |
NM_165509.2 | NP_724535.1 | 4 - 5 | 1531 | 66 | ||
NM_165512.2 | NP_724538.1 | 5 - 6 | 2646 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
