Assay Details
Gene Symbol: Dscam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043020.1 | NP_001036485.1 | 8 - 9 | 2384 | 63 | |
NM_001043057.1 | NP_001036522.1 | 8 - 9 | 2375 | 63 | ||
NM_078925.5 | NP_523649.5 | 8 - 9 | 2381 | 63 | ||
NM_165513.1 | NP_724542.1 | 8 - 9 | 2372 | 63 | ||
NM_165514.1 | NP_724543.1 | 8 - 9 | 2372 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
