Assay Details
Gene Symbol: rgr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169618.2 | NP_001163089.2 | 3 - 4 | 1127 | 67 | |
NM_165611.3 | NP_724690.1 | 2 - 3 | 1028 | 67 | ||
GenBank mRNA | AF353512.1 | - | 2 - 3 | 976 | 67 | |
BT150319.1 | - | 2 - 3 | 999 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
