Assay Details
| Gene Symbol: Jon44E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299274.1 | NP_001286203.1 | 1 | 245 | 91 | |
| NM_165618.2 | NP_724699.1 | 1 | 245 | 91 | ||
| GenBank mRNA | AY095199.1 | - | 1 | 243 | 91 | |
More Information
Set Membership: |
|

Jon44E
Genomic Map