Assay Details
| Gene Symbol: Sec31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299285.1 | NP_001286214.1 | 2 - 3 | 339 | 116 | |
| NM_001299286.1 | NP_001286215.1 | 2 - 3 | 339 | 116 | ||
| NM_136570.4 | NP_610414.1 | 2 - 3 | 339 | 116 | ||
| NM_165630.3 | NP_724716.1 | 2 - 3 | 339 | 116 | ||
More Information
Set Membership: |
|

Sec31
Genomic Map