Assay Details
| Gene Symbol: Sec31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299285.1 | NP_001286214.1 | 7 - 8 | 1822 | 79 | |
| NM_001299286.1 | NP_001286215.1 | 7 - 8 | 1825 | 79 | ||
| NM_136570.4 | NP_610414.1 | 6 - 7 | 1765 | 79 | ||
| NM_165630.3 | NP_724716.1 | 6 - 7 | 1768 | 79 | ||
More Information
Set Membership: |
|

Sec31
Genomic Map