Assay Details
Gene Symbol: Pgi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078939.3 | NP_523663.1 | 2 - 3 | 1221 | 77 | |
NM_165635.2 | NP_724723.1 | 3 - 4 | 1082 | 77 | ||
NM_165636.2 | NP_724724.1 | 2 - 3 | 1038 | 77 | ||
GenBank mRNA | AY051468.1 | - | 2 - 3 | 1212 | 77 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
