Assay Details
Gene Symbol: Pgi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078939.3 | NP_523663.1 | 4 - 5 | 1987 | 84 | |
NM_165635.2 | NP_724723.1 | 5 - 6 | 1848 | 84 | ||
NM_165636.2 | NP_724724.1 | 4 - 5 | 1804 | 84 | ||
GenBank mRNA | AY051468.1 | - | 4 - 5 | 1978 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
