Assay Details
| Gene Symbol: Not1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103772.3 | NP_001097242.1 | 4 - 5 | 2015 | 59 | |
| NM_001103773.3 | NP_001097243.1 | 4 - 5 | 2015 | 59 | ||
| NM_001103774.3 | NP_001097244.1 | 4 - 5 | 1516 | 59 | ||
| NM_001169628.2 | NP_001163099.1 | 4 - 5 | 2015 | 59 | ||
| NM_001299323.1 | NP_001286252.1 | 3 - 4 | 1506 | 59 | ||
| NM_165678.4 | NP_724798.3 | 4 - 5 | 2015 | 59 | ||
More Information
Set Membership: |
|

Not1
Genomic Map