Assay Details
| Gene Symbol: RpL31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136659.3 | NP_610503.1 | 2 - 3 | 363 | 67 | |
| NM_165682.2 | NP_724804.1 | 2 - 3 | 295 | 67 | ||
| NM_165683.2 | NP_724805.1 | 2 - 3 | 308 | 67 | ||
| GenBank mRNA | AY113508.1 | - | 2 - 3 | 300 | 67 | |
More Information
Set Membership: |
|

RpL31
Genomic Map