Assay Details
Gene Symbol: GstT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_165690.4 | NP_724816.3 | 2 - 3 | 292 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
