Assay Details
Gene Symbol: Prosalpha7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_165703.3 | NP_724834.1 | 2 - 3 | 709 | 87 | |
GenBank mRNA | AY069616.1 | - | 2 - 3 | 685 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
